Thulium(III) 2,4-pentanedionate, Powder, REacton™, 99.9% (REO), Alfa Aesar™

Manufacturer: Alfa Aesar 1316403

Quantity 1g

 View more versions of this product

Catalog No. AA1316403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14589-44-7
    Physical Form Powder
    Assay Percent Range 99.9% (REO)
    Molecular Formula C15H21O6Tm
    Formula Weight 466.26
    MDL Number MFCD00058840
    Synonym Thulium(III) acetylacetonate
    Chemical Name or Material Thulium(III) 2, 4-pentanedionate

    Specifications

    Formula Weight 466.26
    Quantity 1g
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.9% (REO)
    Molecular Formula C15H21O6Tm
    MDL Number MFCD00058840
    CAS 14589-44-7
    Synonym Thulium(III) acetylacetonate
    Chemical Name or Material Thulium(III) 2, 4-pentanedionate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit