Learn More
Thulium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Description
Thulium(III) fluoride, anhydrous is used for glass doping and to enhance mid-IR emission in oxyfluoride glass ceramics. It is also used to improve ultraviolet and visible upconversion fluorescence in fluorinated glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2200°C |
Melting Point | 1158°C |
Physical Form | Crystalline |
Quantity | 5 g |
UN Number | UN3288 |
Solubility Information | Insoluble in water |
Formula Weight | 225.93 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
EINECSNumber | 237-353-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.