Alfa Aesar™ Thulium(III) fluoride, ultra dry, 99.99% (REO)

Manufacturer: Alfa Aesar™ 3861506

Quantity 5g

 View more versions of this product

Catalog No. AA3861506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13760-79-7
    Formula Weight 225.93
    MDL Number MFCD00049611
    UN Number UN3288
    Chemical Name or Material Thulium(III) fluoride, Ultra dry

    Specifications

    Formula Weight 225.93
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    Physical Form Powder
    CAS 13760-79-7
    UN Number UN3288
    Melting Point 1158°C
    Density 7.9g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Sensitivity Hygroscopic
    MDL Number MFCD00049611
    Boiling Point 2200°C
    Quantity 5g
    Health Hazard 1 Danger
    TSCA TSCA
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Thulium(III) fluoride, Ultra dry
    Assay 99.99%, (REO)

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit