Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thulium(III) nitrate hydrate, REacton™, 99.99% (REO)

Click to view available options
Quantity:
2g
10g
50g
100641-15-4
MFCD00149875
100641-15-4
MFCD00149875
UN Number UN1477
Solubility Information Soluble in water.
Formula Weight 354.95 (Anhydrous)
Physical Form Wet Crystalline Aggregates
Percent Purity 99.99%
Grade Reacton™
Quantity 2g
Chemical Name or Material Thulium(III) nitrate hydrate
Assay (REO)

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

EINECSNumber : 239-069-9

TSCA : Yes

Recommended Storage : Ambient temperatures