Thulium(III) oxide, 99.99% (REO), REacton™, Alfa Aesar™

Manufacturer: Alfa Aesar 1119909

Quantity 10g

 View more versions of this product

Catalog No. AA1119909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-44-1
    Physical Form Powder
    Assay Percent Range 99.99% (REO)
    Molecular Formula O3Tm2
    Formula Weight 385.87
    MDL Number MFCD00011285

    Specifications

    Formula Weight 385.87
    Quantity 10g
    Color Pale green
    Physical Form Powder
    Assay Percent Range 99.99% (REO)
    Molecular Formula O3Tm2
    MDL Number MFCD00011285
    CAS 12036-44-1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit