Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thulium(III)-sulfate octahydrate, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g,5g
Packaging:
Glass bottle,Glass bottle
13778-40-0
O12S3Tm2·8H2O
770.17
MFCD00149881
GPBYLGMJMIXKMU-UHFFFAOYSA-H
thulium sulfate octahydrate,unii-8zgz55l20j,thulium sulfate octahydrate mi,acmc-20aji8,thulium sulfate tm2 so4 3 octahydrate,thulium sulfate hydrate tm2 so4 3.8h2o,sulfuric acid, thulium 3+ salt 3:2 , octahydrate,thulium sulfate-water 2/3/8,thulium sulfate,,sulfuric acid thulium iii salt octahydrate
71310074
thulium(3+);trisulfate;octahydrate
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3]
13778-40-0
O12S3Tm2·8H2O
770.17
MFCD00149881
GPBYLGMJMIXKMU-UHFFFAOYSA-H
thulium sulfate octahydrate,unii-8zgz55l20j,thulium sulfate octahydrate mi,acmc-20aji8,thulium sulfate tm2 so4 3 octahydrate,thulium sulfate hydrate tm2 so4 3.8h2o,sulfuric acid, thulium 3+ salt 3:2 , octahydrate,thulium sulfate-water 2/3/8,thulium sulfate,,sulfuric acid thulium iii salt octahydrate
71310074
thulium(3+);trisulfate;octahydrate
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3]
for ultra trace analysis
Packaging Glass bottle,Glass bottle
Quantity 5g,5g
Color Green,Green
Assay Percent Range 99.99%
Linear Formula Tm2(SO4)3·8H2O
Merck Index 15, 9548
Formula Weight 770.17
Physical Form Fine Crystalline Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Total Impurity 0.01% max.
Chemical Name or Material Thulium(III)-sulfate octahydrate, 99.99%
Show More Show Less