Learn More
THUMPD1 Mouse anti-Human, Mouse, Non-human primate, Clone: OTI1H6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA526217
Catalog No. PIMA526217
Add to cart
Description
Description
Functions as a tRNA-binding adapter to mediate NAT10-dependent tRNA acetylation (PubMed:25653167).
Specifications
Specifications
THUMPD1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q99J36, Q9NXG2
THUMPD1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
233802, 55623, 696746
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1H6
Unconjugated
THUMPD1
Liquid
THUMPD1
Full length human recombit protein of THUMPD1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Non-human primate