Novus Biologicals™ Thunder-Link oligo Antibody Labeling Kit
Antibody-oligonucleotide conjugates are essential to immuno-PCR. The Thunder-Link™ oligo conjugation system can generate these conjugates more quickly and easily than standard chemistry and methodology.
Manufacturer: Novus Biologicals™ 420-0000
 View more versions of this product
Catalog No. 42-000-00
  Check Availability
Add to Cart
Alert
For Research Use Only
    Specifications
    Quantity 1 Reaction
    Storage Requirements Store at 4°C. Do not freeze.
    Product Type Antibody Labeling Kit
    Features
    • End-user chooses oligo sequence and antibody
    • Oligo sizes (HPLC purified): 10 to 120 bases
    • Amine group positioned at the 5' or 3'
    • Positive control included
    • All necessary components for a successful conjugation are supplied
    • Ideal for immuno-PCR
  • Description & Specifications
Related Products
Product Content Feedback