Learn More
Novus Biologicals™ Thunder-Link oligo Antibody Labeling Kit
Antibody-oligonucleotide conjugates are essential to immuno-PCR. The Thunder-Link™ oligo conjugation system can generate these conjugates more quickly and easily than standard chemistry and methodology.
Manufacturer:  Novus Biologicals™ 4200300
 View more versions of this product
Catalog No. 42-003-00
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
For Research Use Only
Specifications
Description
Features
  • End-user chooses oligo sequence and antibody
  • Oligo sizes (HPLC purified): 10 to 120 bases
  • Amine group positioned at the 5' or 3'
  • Positive control included
  • All necessary components for a successful conjugation are supplied
  • Ideal for immuno-PCR
Description & Specifications
Specifications
3 Reactions
Store at 4°C. Do not freeze.
Antibody Labeling Kit