Learn More
THY1 Mouse anti-Human, Clone: 5.00E+10, Abnova™
Mouse monoclonal antibody raised against human THY1.
Manufacturer:  Abnova Corporation MAB12565
Catalog No. 89-935-007
Add to cart
Description
Description
Specifications
Specifications
THY1
5E10
Mouse monoclonal antibody raised against human THY1.
In PBS, pH 7.2 (0.09% sodium azide).
CD90/FLJ33325
Mouse
IgG1, kappa
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Unconjugated
Flow Cytometry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Liquid
THY1
HEL cells
Affinity purification
RUO
Primary
7070