Learn More
THY1 Mouse anti-Human, Clone: 5.00E+10, Abnova™
Mouse monoclonal antibody raised against human THY1.
Manufacturer:  Abnova Corporation MAB12871
Catalog No. 89-935-008
Add to cart
Description
Description
Specifications
Specifications
THY1
5E10
Mouse monoclonal antibody raised against human THY1.
In PBS, pH 7.4.
CD90/FLJ33325
Mouse
IgG1
100 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Flow Cytometry Immunocytochemistry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined
Liquid
THY1
Human erythroleukemia cell line (HEL).
Protein A purification
RUO
Primary
7070