Alfa Aesar™ Thymidine, 99%

Manufacturer: Alfa Aesar™ A1149306

Quantity 5g

 View more versions of this product

Catalog No. AAA1149306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 50-89-5
    Formula Weight 242.23
    MDL Number MFCD00006537
    Beilstein 89285
    Merck Index 14,9397
    Chemical Name or Material Thymidine

    Specifications

    Formula Weight 242.23
    Physical Form Powder
    TSCA TSCA
    MDL Number MFCD00006537
    Beilstein 89285
    Merck Index 14,9397
    Melting Point 186° to 190°C
    Odor Odorless
    Optical Rotation +19° (c=1 in water)
    CAS 50-89-5
    Quantity 5g
    RTECS Number XP2071000
    Chemical Name or Material Thymidine
    Assay 99%

    Description

    Applications
    Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

    Solubility
    Soluble in water, methanol, hot ethanol, hot acetone, hot ethyl acetate, pyridine, glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit