Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidylate Synthase Mouse anti-Human, Clone: rTYMS/1884, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Thymidylate Synthase
Clone rTYMS/1884
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121764
View Documents
Novus Biologicals
NBP30714520UG
20 μg
Each for $159.82
Add to cart
 
NB121763
View Documents
Novus Biologicals
NBP30714510UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

Thymidylate Synthase Monoclonal antibody specifically detects Thymidylate Synthase in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

Thymidylate Synthase
0.2 mg/ml
Flow Cytometry, ELISA, Immunofluorescence
Purified
RUO
Human
EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422
Recombinant human Thymidylate Synthase protein fragment (around aa 60-174) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
rTYMS/1884
Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control
10 mM PBS with 0.05% BSA
7298
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit