Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidylate Synthase Mouse anti-Human, Clone: SPM453, Novus Biologicals™
Mouse Monoclonal Antibody
$229.00 - $459.00
Specifications
Antigen | Thymidylate Synthase |
---|---|
Clone | SPM453 |
Dilution | Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234390A
|
Novus Biologicals
NBP2343900.1MG |
0.1 mg |
Each for $459.00
|
|
NBP234390F
|
Novus Biologicals
NBP2343900.02MG |
0.02 mg |
Each for $229.00
|
|
NBP234390B
|
Novus Biologicals
NBP2343900.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Thymidylate Synthase Monoclonal specifically detects Thymidylate Synthase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Thymidylate Synthase | |
Western Blot 0.25-0.5ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P04818 | |
7298 | |
Recombinant human thymidylate synthase | |
Primary | |
Store at 4C. | |
36 kDa |
SPM453 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422 | |
TYMS | |
IgG1 κ | |
Protein A purified | |
It recognizes a protein of 36kDa, identified as Thymidylate Synthase (TS) (EC 2.1.1.45). TS converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is also a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title