Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidylate Synthase Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen Thymidylate Synthase
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125120
View Documents
Novus Biologicals
NBP31010925UL
25 μg
Each for $106.19
Add to cart
 
NB125119
View Documents
Novus Biologicals
NBP310109100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

Thymidylate Synthase Polyclonal antibody specifically detects Thymidylate Synthase in Mouse samples. It is validated for Western Blot
Specifications

Specifications

Thymidylate Synthase
Western Blot
Purified
RUO
Mouse
EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422
The immunogen is a synthetic peptide directed towards the C terminal region of mouse Thymidylate Synthase (NP_067263.1). Peptide sequence NHIEPLKIQLQREPRPFPKLKILRKVETIDDFKVEDFQIEGYNPHPTIKM
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control
PBS buffer, 2% sucrose
7298
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit