Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol Blue, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
10g
5g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

76-61-9
C27H30O5S
466.592
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
65565

High quality chemicals for laboratory and specialized industrial use

CAS 76-61-9
Molecular Formula C27H30O5S
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
Molecular Weight (g/mol) 466.592
PubChem CID 65565
Formula Weight 466.6
Packaging Wide Mouth Amber Glass Bottle
Quantity 10g
Chemical Name or Material Thymol Blue
Grade Baker Analyzed™ ACS Reagent
Show More Show Less