Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymol Blue Indicator, 0.4% w/v Aqueous, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Thymol Blue Indicator, 0.4% w/v Aqueous |
CAS | 7732-18-5, 62625-21-2 |
Boiling Point | 100°C |
Synonym | Thymolsulfonphthalein |
Percent Purity | 0.4% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.001 |
Quantity | 120 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction