Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein Complexon, TCI America™

Click to view available options
Quantity:
1g
1913-93-5
C38H44N2O12
720.772
MFCD00001619
AGXCADFSYJNMEG-UHFFFAOYSA-N
112918
2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O
1913-93-5
C38H44N2O12
720.772
MFCD00001619
AGXCADFSYJNMEG-UHFFFAOYSA-N
112918
2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O
Formula Weight 720.77
Physical Form Crystal-Powder at 20°C
Melting Point 255°C
Color White-Yellow
Quantity 1g
Chemical Name or Material Thymolphthalein Complexon