Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroglobulin Mouse anti-Human, Mouse, Rat, Alexa Fluor 750, Clone: 2H11 + 6E1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP234530AF750

Catalog No. NP23430A70


Add to cart

Description

Description

Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human,Mouse,Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

Thyroglobulin
2H11 + 6E1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
AITD3TGN, TDH3, Tg, thyroglobulin
Mouse
IgG
Store at 4°C in the dark.
Monoclonal
Thyroglobulin is a 660kDa dimeric pre-protein with mutiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
TG
Human thyroid follicular cells
0.1 mL
Primary
7038
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit