Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroglobulin Mouse anti-Human, Mouse, Rat, Clone: rTGB/4744, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Thyroglobulin
Clone rTGB/4744
Concentration 0.2 mg/ml
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122175
View Documents
Novus Biologicals
NBP30735110UG
100 μg
Each for $374.34
Add to cart
 
NB122176
View Documents
Novus Biologicals
NBP30735120UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Thyroglobulin
0.2 mg/ml
Flow Cytometry, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Rat
AITD3TGN, TDH3, Tg, thyroglobulin
Human thyroid follicular cells
Monoclonal
Protein A or G purified
rTGB/4744
Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cancer, Cell Biology
10 mM PBS with 0.05% BSA
7038
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit