Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroglobulin Mouse, Clone: TGB04+TGB05, Novus Biologicals™
Mouse Monoclonal Antibody
$229.00 - $459.00
Specifications
Antigen | Thyroglobulin |
---|---|
Clone | TGB04+TGB05 |
Dilution | Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1.0ug/million cells, Immunohistochemistry-Paraffin 0.5-1ug/ml |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234295F
|
Novus Biologicals
NBP2342950.02MG |
0.02 mg |
Each for $229.00
|
|
NBP234295A
|
Novus Biologicals
NBP2342950.1MG |
0.1 mg |
Each for $459.00
|
|
NBP234295B
|
Novus Biologicals
NBP2342950.2MG |
0.2 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Thyroglobulin | |
Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1.0ug/million cells, Immunohistochemistry-Paraffin 0.5-1ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
P01266 | |
7038 | |
Human thyroid follicular cells | |
Primary | |
Store at 4C. | |
660 kDa |
TGB04+TGB05 | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
AITD3TGN, TDH3, Tg, thyroglobulin | |
TG | |
IgG | |
Protein A purified | |
Thyroglobulin is a 660kDa dimeric pre-protein with mutiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title