Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroglobulin Rabbit anti-Human, Clone: TGB/1968R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30735310UG
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Thyroglobulin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
AITD3TGN, TDH3, Tg, thyroglobulin | |
Recombinant full-length human Thyroglobulin protein | |
100 μg | |
Cancer, Cell Biology | |
7038 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
TGB/1968R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title