Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroglobulin Rabbit anti-Human, Clone: TGB/1970R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Thyroglobulin
Clone TGB/1970R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122177
View Documents
Novus Biologicals
NBP30735210UG
100 μg
Each for $406.52
Add to cart
 
NB122178
View Documents
Novus Biologicals
NBP30735220UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Thyroglobulin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
AITD3TGN, TDH3, Tg, thyroglobulin
Recombinant full-length human Thyroglobulin protein
Monoclonal
Protein A or G purified
TGB/1970R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Cell Biology
10 mM PBS with 0.05% BSA
7038
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit