Learn More
Thyroid Peroxidase Mouse anti-Human, Alexa Fluor 488, Clone: TPO34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150812AF488
Catalog No. NBP150812AG
Add to cart
Description
Description
Thyroid Peroxidase Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications
Specifications
Thyroid Peroxidase
TPO34
Western Blot, ELISA
Purified
TPO
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human thyroid peroxidase.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8
Mouse
IgG1
0.25 ml
Lipid and Metabolism
Primary
7173