Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroid Peroxidase Antibody (TPO35), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP150813B
Description
Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for ELISA, Immunoassay, Sandwich ELISA.Specifications
Thyroid Peroxidase | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
TPO | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human thyroid peroxidase. | |
0.25 mL | |
Lipid and Metabolism | |
7173 | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Immunoassay | |
TPO35 | |
ELISA, Immunoassay, Sandwich ELISA | |
EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction