Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thyroid Peroxidase Mouse anti-Human, Clone: MoAb47, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Thyroid Peroxidase |
---|---|
Clone | MoAb47 |
Dilution | Immunohistochemistry-Paraffin 1:10-1:25 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP226475
|
Novus Biologicals
NBP226475 |
0.25 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB019409
|
Novus Biologicals
NBP2264750.05ML |
0.05 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Thyroid Peroxidase Monoclonal antibody specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Thyroid Peroxidase | |
Immunohistochemistry-Paraffin 1:10-1:25 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
TPO | |
IgG1 | |
Tissue culture supernatant |
MoAb47 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Lipid and Metabolism | |
Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide | |
7173 | |
Human Thyroid Peroxidase (hTPO) purified from thyroid microsomes by immunoaffinity chromatography. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title