Learn More
Thyroid receptor-interacting protein 11 Mouse anti-Human, DyLight™ 650, Clone: 10G5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141204C
Catalog No. NBP141204C
Add to cart
Description
Description
Thyroid receptor-interacting protein 11 Monoclonal antibody specifically detects Thyroid receptor-interacting protein 11 in Human samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
Thyroid receptor-interacting protein 11
10G5
Western Blot, Immunocytochemistry/Immunofluorescence
CEV14TR-interacting protein 11, Clonal evolution-related gene on chromosome 14 protein, GMAP-210ACG1A, Golgi-associated microtubule-binding protein 210, Golgi-microtubule-associated protein of 210 kDa, thyroid hormone receptor interactor 11, thyroid receptor-interacting protein 11, TRIP-11, Trip230
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human TRIP230.
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
TRIP11
Amino acids 1099-1372 of Trip230 expressed in E. coli.
0.1 mL
Primary
9321
Human