Learn More
Thyroid Stimulating Hormone Mouse anti-Human, DyLight 488, Clone: Fc1005, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP178579G
Catalog No. NBP178579G
Add to cart
Description
Description
Thyroid Stimulating Hormone Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Thyroid Stimulating Hormone
Fc1005
ELISA
Purified
TSHB
Human Thyroid Stimulating Hormone
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA
Mouse
IgG1
0.1 ml
Cancer
Primary
7252