Learn More
Thyroidreceptor-interactingprotein11 Mouse anti-Human, Janelia Fluor 646, Clone: 10G5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141204JF646
Catalog No. NBP14204646
Add to cart
Description
Description
Thyroidreceptor-interactingprotein11 Monoclonal antibody specifically detects Thyroidreceptor-interactingprotein11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Thyroidreceptor-interactingprotein11
10G5
Immunocytochemistry/Immunofluorescence
Purified
TRIP11
Amino acids 1099-1372 of Trip230 expressed in E. coli.
Protein G purified
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human TRIP230.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CEV14TR-interacting protein 11, Clonal evolution-related gene on chromosome 14 protein, GMAP-210ACG1A, Golgi-associated microtubule-binding protein 210, Golgi-microtubule-associated protein of 210 kDa, thyroid hormone receptor interactor 11, thyroid receptor-interacting protein 11, TRIP-11, Trip230
Mouse
IgG1
0.1mL
Primary
9321
Human