Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIA1 Mouse anti-Human, Clone: TIA1/1313, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14898

Catalog No. 89-034-981


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human TIA1.

  • High specificity
  • High reactivity
Specifications

Specifications

TIA1
TIA1/1313
Mouse monoclonal antibody raised against partial recombinant human TIA1.
In 10 mM PBS.
TIA1
Recombinant protein corresponding to amino acids 279-380 of human TIA1 (exact sequence is proprietary).
Protein A purification
RUO
Primary
7072
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG2b
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)