Tian's Environstar MP Coveralls

Protective apparel manufactured in climate-controlled facility

Manufacturer: Tians International 2168532XL

 View more versions of this product

Catalog No. 19-161-128E

  Check Availability
Add to Cart

    Specifications

    Product Type Elastic wrist, ankles and waist with folded collar
    Certifications/Compliance ISO Class 6, ANSI ISEA 101-1996
    Color White
    For Use With (Application) Protects against petroleum Oils, pesticides, Bleach and Sodium Hydroxide
    Material Polypropylene
    Seam Type Serged
    Size 2X-Large
    Collar Type Folded
    Closure Type Zipper Front
    Waist Style Elastic
    Construction Coated with Microporous Polypropylene
    Ankle Style Elastic
    Disposable Yes
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit