Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tianeptine sodium salt, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
30123-17-2
C21H24ClN2NaO4S
458.933
ZLBSUOGMZDXYKE-UHFFFAOYSA-M
tianeptine sodium,tianeptine sodium salt,stablon,tatinol,coaxil,+-tianeptine monosodium salt,tianeptine sodium, +,sodium 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoate s,s-dioxide,7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoic acid s,s-dioxide sodium salt
23663953
sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-yl)amino]heptanoate
CN1C2=CC=CC=C2C(C3=C(S1(=O)=O)C=C(C=C3)Cl)NCCCCCCC(=O)[O-].[Na+]
This item is not returnable. View return policy
30123-17-2
C21H24ClN2NaO4S
458.933
ZLBSUOGMZDXYKE-UHFFFAOYSA-M
tianeptine sodium,tianeptine sodium salt,stablon,tatinol,coaxil,+-tianeptine monosodium salt,tianeptine sodium, +,sodium 7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoate s,s-dioxide,7-3-chloro-6,11-dihydro-6-methyldibenzo c,f 1,2 thiazepin-11-yl amino heptanoic acid s,s-dioxide sodium salt
23663953
sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-yl)amino]heptanoate
CN1C2=CC=CC=C2C(C3=C(S1(=O)=O)C=C(C=C3)Cl)NCCCCCCC(=O)[O-].[Na+]

Selective facilitator of 5-HT uptake; antidepressant

Selective facilitator of 5-HT uptake in vitro and in vivo. Has no affinity for a wide range of receptors, including 5-HT and dopamine (IC50 > 10 μM) and has no effect on noradrenalin or dopamine uptake. Antidepressant, analgesic and neuroprotective following systemic administration in vivo.
Quantity 10mg
Formula Weight 458.93
Percent Purity >99%
Chemical Name or Material Tianeptine sodium salt

Recommended Storage : Desiccate at Room Temperature