Tianeptine sodium salt, Tocris

Selective facilitator of 5-HT uptake; antidepressant

Manufacturer: R&D Systems 1972/50

Quantity 50mg

 View more versions of this product

Catalog No. 19-725-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C21H24ClN2NaO4S
    Formula Weight 458.93
    Synonym 7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic acid sodium salt
    Chemical Name or Material Tianeptine sodium salt

    Specifications

    Formula Weight 458.93
    Quantity 50mg
    Synonym 7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic acid sodium salt
    Molecular Formula C21H24ClN2NaO4S
    CAS 30123-17-2
    Assay Percent Range >99%
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material Tianeptine sodium salt

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit