Tians Epic Lab Coats

Manufacturer: Tians International 8437852X

 View more versions of this product

Catalog No. 19-138-620E

  Check Availability
Add to Cart

    Specifications

    No. of Pockets 3.0
    Cuff Style Knit
    Material Spunbond Polypropylene
    Size 2X-Large
    For Use With (Application) For general protection and safety
    Closure Type Snap Front
    Color Light Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit