Tians Epic Microporous Coated Lab Coats

Manufacturer: Tians International 8468552XLARGE

 View more versions of this product

Catalog No. 19-161-130E

  Check Availability
Add to Cart

    Specifications

    Material SMS (Spunbound Melt-Blown Synthetic)
    Closure Type Snap
    Construction Virgin Microporous polyethylene film
    Size 2X-Large
    For Use With (Application) Critical and low particulate clean environments
    Disposable Yes
    Certifications/Compliance ANSI, ASTM, ISO Class 6
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit