Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians epic™ High Performance Static Dissipative Frocks

Catalog Number 19161130F
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Size 3X-Large
Material Polypropylene
Color White
Closure Type Snaps
Disposable Single-use
Certifications/Compliance ANSI, ASTM, ISO Class 6
For Use With (Application) Critical and low particulate clean environments
Construction Virgin Microporous polyethylene film
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit