Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians epic™ High Performance Static Dissipative Cleanroom Frocks

Catalog Number 191300519B
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lint-free film coated frock for use in critical, ESD, and low particulate clean environments.

  • Lint-free film coated frock
  • Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas
  • Features include a snap front, elastic wrists
  • Mandarin snap collar, no pockets
  • Fabric is constructed from a blend of polyethylene and polypropylene
  • Finished fabric contains a proprietary additive making it static dissipative
  • For use in ISO 6 Cleanroom Class environments
  • Neatly folded and cleanroom packed
  • 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case
Product Type Controlled Environments Frock
Size Medium
Material Polypropylene, Polyethylene
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Mandarin
Size (English) Chest 11 in.
Size (Metric) Chest 27.9 cm
Certifications/Compliance Class 1 CPSC 1610, IEST-RP-CC003.3
Packaging Type Cleanroom Packed
Length (English) 17 in.
Length (Metric) 43.1 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit