Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ticarcillin disodium and clavulanate potassium (15:1), 50mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
86482-18-0
C23H25N3O11S2
583.583
MFCD26967928
XWMVMWTVLSLJGY-FAJPTIRJSA-N
Timentin
6437075
(2S,5R,6R)-6-[[(2R)-2-carboxy-2-thiophen-3-ylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CSC=C3)C(=O)O)C(=O)O)C.C1C2N(C1=O)C(C(=CCO)O2)C(=O)O
86482-18-0
C23H25N3O11S2
583.583
MFCD26967928
XWMVMWTVLSLJGY-FAJPTIRJSA-N
Timentin
6437075
(2S,5R,6R)-6-[[(2R)-2-carboxy-2-thiophen-3-ylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CSC=C3)C(=O)O)C(=O)O)C.C1C2N(C1=O)C(C(=CCO)O2)C(=O)O
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Ticarcillin disodium and clavulanate potassium (15:1), 50mg/mL in distilled water, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : -30° to -10°C