Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 2WS succinimidyl ester [TF2WS SE] *Superior replacement for FITC*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2349
Tide Fluor™ 2WS (TF2WS) family has the spectral properties similar to those of fluorescein, DyLight 488 and Alexa Fluor 488. Compared to fluorescein probes TF2WS family has better water solubility.
Catalog No. 50-195-3166