Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 3 succinimidyl ester [TF3 SE]*Superior replacement for Cy3* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2271
Tide Fluor™ 3 (TF3) family has the spectral properties essentially identical to those of Cy3. Compared to Cy3 probes TF3 family has much stronger fluorescence and higher photostability.
Catalog No. 50-195-3130