Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 1 alkyne [TF1 alkyne] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2237
TF1 is designed to be a superior fluorophore alternative to EDANS. TF1 has (a). much stronger absorption; (b). much stronger fluorescence intensity; and (3). much less environment-sensitive fluorescence.
Catalog No. 50-195-3112