Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 3WS acid [TF3WS acid] *Superior replacement for Cy3* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2345
Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3. Compared to Cy3 probes TF3WS family has better water solubility.
Catalog No. 50-195-3162