Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 4 maleimide [TF4 maleimide] *Superior replacement for ROX and Texas Red* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2287
TF4 is designed to be a superior fluorophore alternative to ROX and Texas Red®. TF4 has (a). stronger fluorescence intensity; (b). higher conjugation yield; and (c). longer shelf life.
Catalog No. 50-195-3139