Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 5WS alkyne [TF5WS alkyne] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2276
Tide Fluor™ 5WS (TF5WS) family has the spectral properties essentially identical to those of Cy5. Compared to Cy5 probes TF5WS family has much stronger fluorescence and higher photostability.
Catalog No. 50-195-3132