Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 6WS alkyne [TF6WS alkyne] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2303
Tide Fluor™ 6 (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680. Their fluorescence is pH-independent from pH 3 to 11.
Catalog No. 50-195-3148