Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 6WS amine [TF6WS amine] *Superior replacement for Cy5.5* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2292
Tide Fluor™ 6WS (TF6WS) family has the spectral properties similar to those of Cy5.5, IRDye 700 and Alexa Fluor 680. Their fluorescence is pH-independent from pH 3 to 11.
Catalog No. 50-195-3142