Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 7WS azide [TF7WS azide] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2304
Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750. Their fluorescence is pH-independent from pH 3 to 11.
Catalog No. 50-195-3149