Learn More
Aat BioquestSupplier Diversity Partner Tide Fluor™ 8WS amine [TF8WS amine] *Near Infrared Emission* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2336
Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800. Their fluorescence is pH-independent from pH 3 to 11. These characteristics make this new dye family more robust to pH-sensitive assays.
Catalog No. 50-195-3158