Learn More
Aat BioquestSupplier Diversity Partner Tide Quencher™ 1 succinimidyl ester [TQ1 SE] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2199
TQ1 is designed to be a superior quencher alternative to DABCYL. Compared to DABCYL, TQ1 have (a). much stronger absorption; (b). much higher quenching efficiency; and (c).
Catalog No. 50-195-3091