Learn More
Aat BioquestSupplier Diversity Partner Tide Quencher™ 2WS succinimidyl ester [TQ2WS, SE] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2058
TQ2WS is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G. TQ2WS has (a). much stronger absorption; (b). much higher quenching efficiency; and (c).
Catalog No. 50-195-3053